Sodium metabisulfite, for analysis
Sodium metabisulfite, for analysis
Sodium metabisulfite, for analysis
Sodium metabisulfite, for analysis
Thermo Scientific Chemicals
Sodium metabisulfite, for analysis
Reduction reagent | CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA223075000
Price (MYR)
446.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
446.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium metabisulfite
Melting Point150.0°C
CAS7681-57-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Reduction reagent
RUO – Research Use Only