Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Thermo Scientific Chemicals
Sodium hypophosphite monohydrate, for analysis
CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2227900101 kgPlastic bottle
Catalog number FSA222790010
Price (MYR)
1,380.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,380.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium hypophosphite monohydrate
Melting Point90.0°C
CAS10039-56-2
Quantity1 kg
Molecular FormulaH4NaO3P