Potassium nitrite, 97%, for analysis
Potassium nitrite, 97%, for analysis
Potassium nitrite, 97%, for analysis
Potassium nitrite, 97%, for analysis
Thermo Scientific Chemicals
Potassium nitrite, 97%, for analysis
CAS: 7758-09-0 | KNO2 | 85.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA222702500
Price (MYR)
445.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
445.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium nitrite
Melting Point387.0°C
CAS7758-09-0
ColorWhite to Yellow
Health Hazard 1GHS Signal Word: Danger