Tin(II) chloride dihydrate, 98+%
Tin(II) chloride dihydrate, 98+%
Tin(II) chloride dihydrate, 98+%
Tin(II) chloride dihydrate, 98+%
Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 98+%
CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA222665000
Price (MYR)
1,194.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,194.00
BOT
Add to cart
Specifications
Boiling Point652.0°C
Chemical Name or MaterialTin(II) chloride dihydrate
Melting Point37.0°C to 38.0°C
Name Notep.a.
CAS10025-69-1