Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
Thermo Scientific Chemicals
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf
CAS: 14985-18-3 | N2O7Zr | 231.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA222635000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconyl(IV) nitrate hydrate
Name Note99.5%
CAS14985-18-3
ColorWhite
Health Hazard 1GHS Signal Word: Danger