Zinc, granular, 20 mesh
Zinc, granular, 20 mesh
Zinc, granular, 20 mesh
Zinc, granular, 20 mesh
Thermo Scientific Chemicals
Zinc, granular, 20 mesh
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA222605000
Price (MYR)
651.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
651.00
BOT
Add to cart
Specifications
Chemical Name or MaterialZinc
ColorGray
CAS7440-66-6
Melting Point420.0°C
Boiling Point907.0°C
View more
Reduction reagent
RUO – Research Use Only