Tellurium(IV) oxide, 99+%
Tellurium(IV) oxide, 99+%
Tellurium(IV) oxide, 99+%
Tellurium(IV) oxide, 99+%
Tellurium(IV) oxide, 99+%
Thermo Scientific Chemicals
Tellurium(IV) oxide, 99+%
CAS: 7446-07-3 | O2Se | 110.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA222592500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1245°C
Chemical Name or MaterialTellurium(IV) oxide
Melting Point733°C
Name Note99+%
CAS7446-07-3