Tin, granular, 0.1-0.8 mm
Tin, granular, 0.1-0.8 mm
Tin, granular, 0.1-0.8 mm
Tin, granular, 0.1-0.8 mm
Thermo Scientific Chemicals
Tin, granular, 0.1-0.8 mm
CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA222575000
Price (MYR)
911.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
911.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTin
ColorGray
Name Notep.a.
CAS7440-31-5
Melting Point231.9°C
View more
Reduction reagent
RUO – Research Use Only