Barium sulfate, 98%, extra pure
Barium sulfate, 98%, extra pure
Barium sulfate, 98%, extra pure
Barium sulfate, 98%, extra pure
Thermo Scientific Chemicals
Barium sulfate, 98%, extra pure
Barium sulfate, 98%, BaO4S, CAS Number-7727-43-7, barite, baritop, barium sulfate, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast, 25kg, Passes test, < 0.3%, 97.5 to 100.5%, White, 4.5000g/mL, 231-784-4, 233.4, Extra Pure, 10ppm max., TZCXTZWJZNENPQ-UHFFFAOYSA-L | CAS: 7727-43-7 | BaO4S | 233.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA222510050
Price (MYR)
1,281.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,281.00
EA
Add to cart
Specifications
Chemical Name or MaterialBarium sulfate
Melting Point1580.0°C
CAS7727-43-7
ColorWhite
Physical FormPowder