Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Thermo Scientific Chemicals
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA222220250
Price (MYR)
1,898.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,898.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiphenylamine-4-sulfonic acid, sodium salt
Name Notep.a.
CAS6152-67-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
View more
Indicator
RUO – Research Use Only