3-Iodoanisole, 97%
3-Iodoanisole, 97%
3-Iodoanisole, 97%
Thermo Scientific Chemicals
3-Iodoanisole, 97%
CAS: 766-85-8 | C7H7IO | 234.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA222020250
Price (MYR)
542.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
542.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Iodoanisole
CAS766-85-8
CAS Min %96.0
CAS Max %100.0
ColorYellow