Specifications
Chemical Name or MaterialBromoacetonitrile
Name Note97%
CAS590-17-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.