N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
Thermo Scientific Chemicals
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
CAS: 24589-78-4 | C6H12F3NOSi | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number FSA221581000
Price (MYR)
4,892.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,892.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Methyl-N-(trimethylsilyl)trifluoroacetamide
CAS24589-78-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Alkylation reagent, Silylation reagent
RUO – Research Use Only