N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%
Thermo Scientific Chemicals
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%
CAS: 77377-52-7 | C9H18F3NOSi | 241.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA221540050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide
CAS77377-52-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Alkylation reagent, Silylation reagent
RUO – Research Use Only