View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenetole
Name Note99%
CAS103-73-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Flammable liquid and vapour.