1-Heptadecanol, 97%
1-Heptadecanol, 97%
1-Heptadecanol, 97%
Thermo Scientific Chemicals
1-Heptadecanol, 97%
CAS: 1454-85-9 | C17H36O | 256.474 g/mol
Have Questions?
Catalog NumberQuantity
FSA2212300505 g
Catalog number FSA221230050
Price (MYR)
390.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
390.00
EA
Add to cart
Specifications
Chemical Name or Material1-Heptadecanol
CAS1454-85-9
Melting Point51.0°C to 56.0°C
ColorWhite
EINECS Number215-932-5