Hexacosane, 99%
Hexacosane, 99%
Hexacosane, 99%
Thermo Scientific Chemicals
Hexacosane, 99%
CAS: 630-01-3 | C26H54 | 366.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA221220050
Price (MYR)
598.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
598.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexacosane
Name Note99%
CAS630-01-3
Melting Point56°C to 59°C