2-Chloroethyl isocyanate, 98%
2-Chloroethyl isocyanate, 98%
2-Chloroethyl isocyanate, 98%
2-Chloroethyl isocyanate, 98%
Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%
CAS: 1943-83-5 | C3H4ClNO | 105.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA221190250
Price (MYR)
1,195.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,195.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chloroethyl isocyanate
Name Note98%
CAS1943-83-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger