N-Phenyl-p-phenylenediamine, 98%
N-Phenyl-p-phenylenediamine, 98%
N-Phenyl-p-phenylenediamine, 98%
N-Phenyl-p-phenylenediamine, 98%
Thermo Scientific Chemicals
N-Phenyl-p-phenylenediamine, 98%
CAS: 101-54-2 | C12H12N2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA221001000
Price (MYR)
434.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
434.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Phenyl-p-phenylenediamine
Name Note98%
CAS101-54-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning