Glycine, 99+%, for analysis
Glycine, 99+%, for analysis
Glycine, 99+%, for analysis
Glycine, 99+%, for analysis
Thermo Scientific Chemicals
Glycine, 99+%, for analysis
Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins. | CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA220912500
Price (MYR)
286.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
286.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycine
Ammonium200ppm max.
CAS56-40-6
Arsenic (As)1ppm max.
ColorWhite
View more
General description
  • Glycine is an essential acid that can be used for synthesis of creatine, purines and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity
Applications
  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
RUO – Research Use Only