Acetic acid, sodium salt, 99+%, for analysis, anhydrous
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change view
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA220892500
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
Name Notep.a.
pH7.5 to 9.2 (5% soln. at 20°C)
CAS127-09-3
Melting Point324.0°C