1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
1,2-Propanediol, 99+%, for analysis
Thermo Scientific Chemicals
1,2-Propanediol, 99+%, for analysis
CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA220872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Propanediol
CAS57-55-6
CAS Min %99.0
CAS Max %100.0
Melting Point-60.0°C