Gallic acid monohydrate
Gallic acid monohydrate
Gallic acid monohydrate
Gallic acid monohydrate
Thermo Scientific Chemicals
Gallic acid monohydrate
CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA220765000
Price (MYR)
995.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
995.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGallic acid monohydrate
Name Notec.p.
CAS5995-86-8
Melting Point251.0°C
Health Hazard 1Warning