Dimethyl malonate, 99+%
Dimethyl malonate, 99+%
Dimethyl malonate, 99+%
Dimethyl malonate, 99+%
Thermo Scientific Chemicals
Dimethyl malonate, 99+%
CAS: 108-59-8 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA220702500
Price (MYR)
279.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
279.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl malonate
Name Note99+%
CAS108-59-8
Melting Point-62.0°C