Propylamine, 99+%, extra pure
Propylamine, 99+%, extra pure
Propylamine, 99+%, extra pure
Propylamine, 99+%, extra pure
Thermo Scientific Chemicals
Propylamine, 99+%, extra pure
CAS: 107-10-8 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA220615000
Price (MYR)
448.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropylamine
Name Note99+%
CAS107-10-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.