Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Thermo Scientific Chemicals
Styrene, 99.5%, for analysis, stabilized
CAS: 100-42-5 | C8H8 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA220532500
Price (MYR)
261.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
261.00
BOT
Add to cart
Specifications
Chemical Name or MaterialStyrene
Name Notep.a.
CAS100-42-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
May be fatal if swallowed and enters airways.<br/