Methyl salicylate, 99%
Methyl salicylate, 99%
Methyl salicylate, 99%
Methyl salicylate, 99%
Thermo Scientific Chemicals
Methyl salicylate, 99%
CAS: 119-36-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA220495000
Price (MYR)
401.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
401.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl salicylate
CAS119-36-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.