Ethyl acetoacetate, 99+%, extra pure
Ethyl acetoacetate, 99+%, extra pure
Ethyl acetoacetate, 99+%, extra pure
Ethyl acetoacetate, 99+%, extra pure
Thermo Scientific Chemicals
Ethyl acetoacetate, 99+%, extra pure
CAS: 141-97-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA220401000
Price (MYR)
203.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
203.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetoacetate
Name Notep.a.
CAS141-97-9
Melting Point-45°C
Density1.0300g/mL