Citric acid, 99.5%, for analysis, anhydrous
Citric acid, 99.5%, for analysis, anhydrous
Citric acid, 99.5%, for analysis, anhydrous
Citric acid, 99.5%, for analysis, anhydrous
Thermo Scientific Chemicals
Citric acid, 99.5%, for analysis, anhydrous
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA220345000
Price (MYR)
510.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
510.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitric acid
CAS77-92-9
Melting Point153.0°C
Health Hazard 1Warning
ColorWhite
View more
Biological buffer for analysis
RUO – Research Use Only