Chloroacetic acid, 99+%
Chloroacetic acid, 99+%
Chloroacetic acid, 99+%
Chloroacetic acid, 99+%
Thermo Scientific Chemicals
Chloroacetic acid, 99+%
CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA220321000
Price (MYR)
202.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
202.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChloroacetic acid
Name Notep.a.
CAS79-11-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.