p-Xylene, 99+%, extra pure
p-Xylene, 99+%, extra pure
p-Xylene, 99+%, extra pure
p-Xylene, 99+%, extra pure
Thermo Scientific Chemicals
p-Xylene, 99+%, extra pure
CAS: 106-42-3 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA220210010
Price (MYR)
595.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
595.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Xylene
Melting Point13.0°C
Name NoteExtra Pure
CAS106-42-3
Health Hazard 1Danger