Thiourea, 99+%, for analysis
Thiourea, 99+%, for analysis
Thiourea, 99+%, for analysis
Thiourea, 99+%, for analysis
Thermo Scientific Chemicals
Thiourea, 99+%, for analysis
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA220052500
Price (MYR)
301.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
301.00
BOT
Add to cart
Specifications
Chemical Name or MaterialThiourea
Name NoteFor analysis
CAS62-56-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.
View more
For analysis
RUO – Research Use Only