Acetic acid, ammonium salt, 98+%, for HPLC
Acetic acid, ammonium salt, 98+%, for HPLC
Acetic acid, ammonium salt, 98+%, for HPLC
Acetic acid, ammonium salt, 98+%, for HPLC
Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98+%, for HPLC
CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Plastic bottle
Catalog number FSA219990500
Price (MYR)
509.00
EA
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
509.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, ammonium salt
CAS631-61-8
Melting Point114.0°C
Molecular FormulaC2H7NO2
Linear FormulaCH3CO2NH4
View more
For HPLC
RUO – Research Use Only