2,6-Di-tert-butyl-4-methylphenol, 99.8%
2,6-Di-tert-butyl-4-methylphenol, 99.8%
2,6-Di-tert-butyl-4-methylphenol, 99.8%
2,6-Di-tert-butyl-4-methylphenol, 99.8%
Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methylphenol, 99.8%
CAS: 128-37-0 | C15H24O | 220.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA219832500
Price (MYR)
294.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
294.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Di-tert-butyl-4-methylphenol
Name Notep.a.
CAS128-37-0
CAS Min %99.75
Health Hazard 1GHS Signal Word: Warning