Diisopropylamine, 99+%, Thermo Scientific Chemicals
Diisopropylamine, 99+%, Thermo Scientific Chemicals
Diisopropylamine, 99+%, Thermo Scientific Chemicals
Diisopropylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diisopropylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA2194300101 LGlass bottle
Catalog number FSA219430010
Price (MYR)
704.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
704.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiisopropylamine
Name Note99+%
CAS108-18-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.