N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
Thermo Scientific Chemicals
N,N-Dimethylethylamine, 99%
CAS: 598-56-1 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA219292500
Price (MYR)
590.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
590.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylethylamine
Name Note99%
CAS598-56-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger