Metaphosphoric acid, stabilized
Metaphosphoric acid, stabilized
Metaphosphoric acid, stabilized
Metaphosphoric acid, stabilized
Thermo Scientific Chemicals
Metaphosphoric acid, stabilized
Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max. | CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA219225000
Price (MYR)
1,699.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,699.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMetaphosphoric acid
CAS37267-86-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
CAS50813-16-6