Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Thermo Scientific Chemicals
Chromium(III) nitrate nonahydrate, 99%
CAS: 7789-02-8 | CrN3O9·9H2O | 400.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
5 kg
Packaging:
Plastic bottle
Catalog number FSA219200010
Price (MYR)
1,262.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,262.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChromium(III) nitrate nonahydrate
Melting Point60.0°C
Name Note99%
CAS7789-02-8
ColorBlack to Purple