Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Thermo Scientific Chemicals
Drierite™, with indicator, 8 mesh
Drierite, CaO4S, CAS Number-7778-18-9, 7646-79-9, 5kg, 39.09%, 0.68%, CHEBI:31346, 3.25%, Blue, 01,107, 136.13, GHS Signal Word: Danger | CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Packaging:
Glass Bottle
Catalog number FSA219095000
Price (MYR)
605.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
605.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDrierite™
ColorBlue
Physical FormGranular
Name NoteWith indicator, 8 Mesh
CAS7778-18-9
View more
Adsorbent, Indicator
RUO – Research Use Only