Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Drierite™, without indicator, 8 mesh
Thermo Scientific Chemicals
Drierite™, without indicator, 8 mesh
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass Bottle
Catalog number FSA219055000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDrierite™
Melting Point1450.0°C
Physical FormGranular
CAS7778-18-9
Health Hazard 1
View more
Adsorbent, Indicator
RUO – Research Use Only