Trolox™, 97%
Trolox™, 97%
Trolox™, 97%
Thermo Scientific Chemicals
Trolox™, 97%
CAS: 53188-07-1 | C14H18O4 | 250.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA218940250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTrolox*
Name Note97%
CAS53188-07-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: