3-Methyl-2-butanone, 98%
3-Methyl-2-butanone, 98%
3-Methyl-2-butanone, 98%
3-Methyl-2-butanone, 98%
Thermo Scientific Chemicals
3-Methyl-2-butanone, 98%
CAS: 563-80-4 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA218772500
Price (MYR)
415.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
415.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Methyl-2-butanone
CAS563-80-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0