Cesium sulfate, 99+%, pure
Cesium sulfate, 99+%, pure
Cesium sulfate, 99+%, pure
Cesium sulfate, 99+%, pure
Thermo Scientific Chemicals
Cesium sulfate, 99+%, pure
CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA218702500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1900.0°C
Chemical Name or MaterialCesium sulfate
Melting Point1005.0°C
Name Note99+%
CAS10294-54-9