Cerium(III) nitrate hexahydrate, 99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Thermo Scientific Chemicals
Cerium(III) nitrate hexahydrate, 99.5%
CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA218695000
Price (MYR)
1,590.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,590.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCerium(III) nitrate hexahydrate
Name Note99.5%
CAS10294-41-4
ColorColorless to White
Health Hazard 1GHS Signal Word: Danger