1,3-Diiodopropane, 99%, stabilized
1,3-Diiodopropane, 99%, stabilized
1,3-Diiodopropane, 99%, stabilized
1,3-Diiodopropane, 99%, stabilized
Thermo Scientific Chemicals
1,3-Diiodopropane, 99%, stabilized
CAS: 627-31-6 | C3H6I2 | 295.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA218600250
Price (MYR)
1,033.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,033.00
BOT
Add to cart
Specifications
Chemical Name or Material1,3-Diiodopropane
Name Note99%
CAS627-31-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning