Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Ruthenium(IV) oxide, 99.5+%, anhydrous
Thermo Scientific Chemicals
Ruthenium(IV) oxide, 99.5+%, anhydrous
CAS: 12036-10-1 | O2Ru | 133.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA218340050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRuthenium(IV) oxide
Name Note99.5+%
CAS12036-10-1
ColorBlack
Health Hazard 1GHS Signal Word: Warning