Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
CAS: 13494-98-9 | N3O96H2O | 383.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA218265000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialYttrium(III) nitrate hexahydrate
Name Note99.9%
CAS13494-98-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
For ultra trace analysis
RUO – Research Use Only