Bis(triphenylphosphine)nickel(II)chloride, 98%
Bis(triphenylphosphine)nickel(II)chloride, 98%
Bis(triphenylphosphine)nickel(II)chloride, 98%
Bis(triphenylphosphine)nickel(II)chloride, 98%
Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%
CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA217500500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBis(triphenylphosphine)nickel(II)chloride
Name Note98%
CAS14264-16-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if inhaled.
May cause cancer.