n-Pentane, 99+%, extra pure, anhydrous
n-Pentane, 99+%, extra pure, anhydrous
n-Pentane, 99+%, extra pure, anhydrous
n-Pentane, 99+%, extra pure, anhydrous
Thermo Scientific Chemicals
n-Pentane, 99+%, extra pure, anhydrous
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA217240025
Price (MYR)
1,087.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,087.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Pentane
Melting Point-130.0°C
Name Notep.a.
CAS109-66-0
CAS Min %99.0