Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA217115000
Price (MYR)
330.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
330.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt trihydrate
Name Notep.a.
pH7.5 to 9.0 (5 % at 20°C)
CAS127-09-3
Melting Point58.0°C
View more
Biological buffer
RUO – Research Use Only